Learn More
Grainger Sq. End Mill Single End Carb 10.00mm
Supplier:  Grainger 14G090
Encompass_Preferred
The Micro 100 Square End Mill Center Cutting 14G090 is a square end milling tool made by Micro 100. It has a 1/4" shank diameter, 1/2" cutting diameter, and 1-1/2" overall length. The end mill features a TiAlN coating for increased tool life and heat resistance. It is designed for center cutting in low carbon steels at speeds up to 6,000 RPM with .002-.005" chip load per tooth. This is a general purpose square end mill suitable for slotting, side milling and contouring operations.
Catalog No. 50-275-3430