Learn More
Grainger Sq. End Mill Single End Carb 15/64
Supplier:  Grainger 14M422
Encompass_Preferred
The Grainger Approved Square End Mill Single End is a high-speed steel end mill with a square end for milling slots, keyways, and pockets. It has four flutes and a right-hand spiral. The overall length is 3 inches and the shank diameter is 1/2 inch. The square end measures 5/8 inch x 5/8 inch. This end mill is uncoated and made in the USA. It can machine low and medium carbon steels at surface speeds up to 120 SFM when used with rigid setups.
Catalog No. 50-275-1223