Learn More
Grainger Sq. End Mill Single End Carb 1/2
Supplier:  Grainger 14M521
Encompass_Preferred
The Grainger Approved Square End Mill Single End Carbide is a milling cutter with a square end for machining slots, shoulders, and angles. It has a 1/2" shank diameter, 1" overall length, and 1/2" cutting diameter. The end mill is made of solid carbide for wear resistance and has 2 flutes for efficient chip removal. It can machine steels, cast iron, aluminum, and other non-ferrous metals at high speeds and feeds. This is a high quality carbide end mill for light to medium duty milling applications.
Catalog No. 50-275-1489