Learn More
Grainger Sq. End Mill Single End Carb 1/32
Supplier:  Grainger 14M341
Encompass_Preferred
The Micro 100 Square End Mill Center Cutting is a high-performance end mill made in the USA by Micro 100. It features a square end design for milling 90 degree shoulders and angles. The center cutting geometry allows plunge cuts into the middle of a workpiece. This tool has 4 flutes and is made of solid carbide for strength and durability when milling steel, aluminum, and other materials. The 1/4 inch shank diameter fits standard tool holders. It comes uncoated for general purpose milling applications. Overall this is a versatile square end mill for shoulder milling, slotting, side milling, and contouring operations.
Catalog No. 50-275-0061