Learn More
Grainger Sq. End Mill Single End Carb 1.80mm
Supplier:  Grainger 14K568
Encompass_Preferred
The Grainger Approved Square End Mill Single End is a high-speed steel end mill with a square end for milling slots, keyways, and pockets. It has a straight shank and is uncoated with an overall length of 3 inches and a shank diameter of 1/2 inch. The number of flutes is 2 and it has a right hand spiral. The end mill has an American Standard taper shank and a square end style. The primary application is slotting.
Catalog No. 50-274-9591