Learn More
Grainger Sq. End Mill Single End Carb 13/64
Supplier:  Grainger 14M408
Encompass_Preferred
The Micro 100 Square End Mill Center Cutting is a high-performance end mill designed for milling a wide range of materials. This 1/4" diameter, 4 flute end mill features a square end for milling 90 degree shoulders and corners. It has a center cutting design that allows plunge cutting and its TiAlN coating provides heat and wear resistance. Made in the USA out of solid carbide, this end mill is rigid and durable for increased tool life. With a neck relief design, it reduces heat buildup and allows for increased feed rates. The Micro 100 Square End Mill Center Cutting is well-suited for slotting, side milling and facing in steels, cast iron, aluminum and other non-ferrous metals.
Catalog No. 50-275-0051