Learn More
Grainger Boltless Shelving 18x84x60in AdOn
Supplier:  Grainger 14L155
Encompass_Preferred
The Boltonless Shelving by Tennsco is an 18" deep x 84" high x 60" wide steel shelving unit with 5 adjustable shelves. It has a weight capacity of 800 lbs per shelf and 4000 lbs total. The unit assembles without nuts, bolts, or clips using the company's boltless design with welded frames. It can be used for industrial, retail, or office storage needs. The gray powder coat finish resists chipping and scratching.
Catalog No. 50-275-1370