Learn More
Grainger Sq. End Mill Single End Carb 3/32
Supplier:  Grainger 14T295
Encompass_Preferred
The Grainger Approved Square End Mill Single End is a high-speed steel end mill with a square end for milling slots, keyways, and pockets. It has 4 flutes and a right-hand spiral. The overall length is 3 inches and the shank diameter is 1/2 inch. The mill diameter is 1 inch with a 1/8 inch shank diameter. It has an American National standard taper shank and can be used on materials like low carbon steel, brass, aluminum and plastic. This end mill is good for light duty milling applications.
Catalog No. 50-274-8861