Learn More
Grainger Sq. End Mill Single End Carb 3/16
Supplier:  Grainger 14F964
Encompass_Preferred
The Micro 100 Square End Mill Center Cutting 14F964 is a square end milling cutter with a center cutting edge. It has a 1/2" shank diameter and a 1/2" cutting diameter. The overall length is 2-1/2". It is made of solid carbide for wear resistance and has 2 flutes. The square end allows milling of shoulders and slots. The center cutting edge allows plunging. It can be run at speeds up to 325 SFM. This is a versatile and durable mill for a wide range of materials.
Catalog No. 50-275-3581