Learn More
Grainger Sq. End Mill Single End Carb 3/32
Supplier:  Grainger 14T296
Encompass_Preferred
The Micro 100 Square End Mill Carbide is a high-performance end mill for milling a variety of materials. It features a square end design for milling 90-degree shoulders and angles. The end mill has four flutes and is made of solid carbide with TiAlN coating for wear resistance and heat resistance. It has a 1/2" shank diameter and is available in multiple diameters from 1/8" to 1/2". The square end mill is ideal for slotting, side milling, and facing on materials like steel, stainless steel, cast iron, aluminum, titanium, and plastics. It can achieve precise right angle shoulders in corner milling applications.
Catalog No. 50-274-9483