Learn More
Grainger Sq. End Mill Single End Carb 6.00mm
Supplier:  Grainger 14M690
Encompass_Preferred
The Grainger Approved Square End Mill Single End is a high-speed steel end mill with a square end for milling slots, keyways, and pockets. It has a 1/2" shank diameter, a 1-1/2" overall length, and a 1/2" cutter diameter. The number of flutes is 2. This end mill is uncoated and made in the USA. The primary applications are slotting, side milling, and pocketing in low carbon steels at surface speeds up to 115 SFM when used with rigid setups.
Catalog No. 50-274-9605