Learn More
Grainger Sq. End Mill Single End Carb 6.00mm
Supplier:  Grainger 14T213
Encompass_Preferred
The Micro 100 Square End Mill Center Cutting is a high-performance end mill designed for milling a wide range of materials. This 1/4" diameter, 4 flute end mill has a square end for milling 90 degree shoulders and slots in aluminum, steel, stainless steel, cast iron, titanium, plastic and other materials. It features a TiAlN coating for heat and wear resistance during high speed machining. The center cutting design allows plunge cutting and drilling. Recommended speed is 25,000 RPM with feed rates from 0.001-0.004 IPR based on material. Made in USA with a limited lifetime warranty.
Catalog No. 50-275-1592