Learn More
Grainger Sq. End Mill Single End Carb 6.00mm
Supplier:  Grainger 14G079
Encompass_Preferred
The Grainger Approved Square End Mill Single End is a high-speed steel end mill with a square end for milling slots, keyways, and pockets. It has four flutes and a right-hand spiral. The overall length is 3 inches and the shank diameter is 0.5 inches. This end mill is uncoated and made in the USA. It can be used on steel, stainless steel, cast iron, aluminum, brass, bronze, and plastic. The maximum RPM is 2700.
Catalog No. 50-275-1544