Learn More
Grainger Sq. End Mill Single End Carb 7/64
Supplier:  Grainger 14M372
Encompass_Preferred
The Micro 100 Square End Mill Center Cutting 14M721 is a high-performance end mill designed for milling a wide range of materials. This 1/4" diameter, 4 flute end mill features a square end for milling 90 degree shoulders and corners in steels, tool steels, stainless steels, cast iron, aluminum, copper, brass, bronze, plastics and other materials. The center cutting design allows plunge cuts into the middle of a workpiece. The TiAlN coating provides heat resistance and lubricity. Overall, this is a versatile square end mill well-suited for shoulder milling, slotting, side milling and contouring operations.
Catalog No. 50-274-9990