Learn More
Grainger Sq. End Mill Single End Carb 7/32
Supplier:  Grainger 14M416
Encompass_Preferred
The Micro 100 Square End Mill Center Cutting is a high performance square end mill designed for milling a wide range of materials. It features a center cutting design for plunge milling and slotting operations. The end mill has a square shape with four flutes and is made of solid carbide with TiAlN coating for wear resistance and heat resistance. It has a 1/2" shank diameter and is available in multiple sizes from 1/8" to 1/2" cutting diameters. The square end mill can achieve precise 90 degree shoulders in corner milling applications. Its center cutting geometry allows for plunge cuts into the middle of a workpiece. This versatile end mill is rigid and durable for increased tool life.
Catalog No. 50-275-1599