Learn More
Grainger Sq. End Mill Single End Carb 9/64
Supplier:  Grainger 14T314
Encompass_Preferred
The Grainger Approved Square End Mill Single End is a high-speed steel end mill with a square end for milling slots, keyways, and pockets. It has a 1/2" shank diameter, 1-1/2" overall length, and a 1/4" cutting diameter. The end mill has 4 flutes and a right-hand spiral. It is designed for use on steel, stainless steel, cast iron, aluminum, brass, bronze, and plastic. The end mill provides accurate cuts and smooth finish. It is manufactured in the USA to tight tolerances.
Catalog No. 50-275-0941