Learn More
Chemglass Life Sciences Media Bottle Assembly, PETG, 1,000 mL, 2-Port, Sterile, Single-Use
SDP
Supplier:  Chemglass Life Sciences CLS1168X1000
Encompass
Single-use 1,000 mL PETG media bottle assembly for sterile liquid transfer and containment. The assembly includes a 2-port closure with a 30 mm, 0.22 μm vent filter, 24 in of weldable TPE tubing with a plugged end and a 1/4 in pinch clamp; units are double poly-bagged and gamma irradiated. Typically packaged in packs of 5.
  • High-clarity PETG bottle with molded graduations.
  • 2-port closure with 30 mm, 0.22 μm vent filter.
  • 24 in of weldable TPE tubing with a plugged end.
  • V-notched dip tube extends to the bottom for complete drainability.
  • Stopper design includes strain relief to prevent tubing kinking.
  • Manufactured and packaged in a Class 7 cleanroom.
  • Gamma irradiated and double poly-bagged for sterility assurance.
  • Materials meet USP Class VI and are animal derived component free.
Catalog No. NC3481769