Learn More
TARGETMOL CHEMICALS INC SQ109 5MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T169255MG
Encompass_Preferred
Also available in 1 mL, 1 mg, 10 mg, 25 mg, 50 mg, 100 mg, 500 mg and bulk. Please contact Fisher for quotes. SQ109 (NSC-722041) is an effective inhibitor of the trypomastigote form of the parasite (IC50 = 50 nM). SQ109 is an antitubercular agent and targets MmpL3. Purity 99.3%
Catalog No. 50-287-7400