Learn More
Analytical Sales & Services Square Well Reservoir Plate, 24 Wells, 10 mL Capacity, Clear Polypropylene
Supplier:  Analytical Sales & Services 24727
Encompass
The SQR WELL RESERVOIR PLATE is a high-capacity, square well microplate designed for laboratory sample management. Each of the 24 wells holds 10 mL, maximizing sample volume while ensuring chemical compatibility. Made from clear polypropylene, it allows easy sample visibility and is compatible with robotic systems for efficient workflow. Ideal for various applications including reactions and automated liquid handling, this 25-pack plate enhances your research efficiency.
Catalog No. NC0640066