Learn More
TARGETMOL CHEMICALS INC Squalene 200MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T4749200MG
Encompass_Preferred
Also available in 1 mL, 10 mg, 25 mg, 50 mg, 100 mg, 500 mg and bulk. Please contact Fisher for quotes. Squalene (AddaVax) is a natural organic compound originally obtained for commercial purposes primarily from shark liver oil, though there are botanical sources as well, including rice bran, wheat germ, and olives. All higher organisms produce squalene, including humans. Squalene is an intermediate product in the synthesis of cholesterol and shows several pharmacological properties such as hypolipidemic, hepatoprotective, cardioprotective, antioxidant, and antitoxicant activity. Purity 99.33%
Catalog No. 50-287-9979