Learn More
Apexbio Technology LLC SR 11237 146670-40-8 10mg
SDP
Supplier:  Apexbio Technology LLC B739810
Encompass_Preferred
SR 11237 (146670-40-8) is a small-molecule agonist targeting the retinoid X receptor (RXR) It is designed to selectively activate RXR thereby modulating RXR-mediated transcriptional regulation SR 11237 exerts its biological activity primarily through specific binding to RXR facilitating RXR-RXR homodimer formation and activating genes via RXR response elements Based on these pharmacological properties SR 11237 holds research potential in the investigation of RXR-dependent transcription nuclear receptor signaling pathways and therapeutic strategies related to RXR-associated diseases
Catalog No. 50-000-91056