Learn More
Medchemexpress LLC N-((6,7-difluoro-1H-benzo[d]imidazol-2-yl)methyl)-9-(3-fluorophenyl)-2-morpholino-9H-purin-6-amine | 1454585-06-8 | MFCD30187527 | 99.7% | 480.45 | C23H19F3N8O | 100 MG
SDP
Supplier:  Medchemexpress LLC HY100011100MG
Encompass_Preferred
SR-3029 is a small-molecule, ATP-competitive inhibitor of casein kinase 1 delta (CK1δ) and casein kinase 1 epsilon (CK1ε), intended for research use in biochemical and cell-based studies. It exhibits potent enzyme inhibition and is provided in formats suitable for assay and formulation work.
  • Potent ATP-competitive inhibition of CK1δ and CK1ε.
  • Low-nanomolar activity against CK1δ (IC50 44 nM).
  • Sub-micromolar activity against CK1ε (IC50 260 nM).
  • Ki of 97 nM for both CK1δ and CK1ε.
  • Chemical formula C23H19F3N8O and molecular weight 480.45.
  • High purity (99.7%) suitable for research applications.
  • Available as solid and as DMSO solution for experimental flexibility.
Catalog No. 50-002-15442
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.