Learn More
Medchemexpress LLC MRT68921 | 1190379-70-4 | MFCD30533276 | 99.7% | 434.58 g/mol | C25H34N6O | 25 MG
SDP
Supplier:  Medchemexpress LLC HY10000625MG
Encompass_Preferred
MRT68921 is a potent NUAK1/ULK1 dual kinase inhibitor used in research to block autophagy, inhibit tumor cell proliferation, and study oxidative stress-mediated apoptosis. It is provided in small milligram quantities for laboratory research and is available in multiple salt forms.
  • Potent ULK1 and ULK2 inhibition with low-nanomolar IC50 values.
  • Used to block autophagy and induce apoptosis in cell models.
  • Available in multiple salt forms to support experimental needs.
  • High reported purity suitable for research applications.
  • Supplied in milligram-scale quantities for preclinical studies.
  • Recommended cold storage to maintain compound stability.
Catalog No. 50-002-15790