Learn More
Medchemexpress LLC N-[(4,5-difluoro-1H-benzimidazol-2-yl)methyl]-9-(3-fluorophenyl)-2-morpholin-4-ylpurin-6-amine | 1454585-06-8 | MFCD30187527 | 99.7% | 480.4 g/mol | C23H19F3N8O | 1 ML
Supplier: Medchemexpress LLC HY10001110MM 1ML
SR-3029 is a potent, ATP-competitive inhibitor of casein kinase 1 delta (CK1δ) and epsilon (CK1ε) for biochemical and cellular research. It exhibits nanomolar activity and is supplied as a 10 mM solution in DMSO for laboratory use.
- Potent ATP-competitive inhibitor of CK1δ and CK1ε.
- IC50: CK1δ 44 nM; CK1ε 260 nM.
- Ki 97 nM for both kinases.
- Provided as a 10 mM solution in DMSO (1 mL).
- High purity (>99.7%) suitable for research applications.
- Useful for biochemical assays and cellular kinase studies.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.