Learn More
Medchemexpress LLC N-[(6,7-difluoro-1h-benzimidazol-2-yl)methyl]-9-(3-fluorophenyl)-2-(4-morpholinyl)-9h-purin-6-amine | 1454585-06-8 | MFCD30187527 | 99.7% | 480.45 g·mol⁻¹ | C23H19F3N8O | 25 MG
SDP
Supplier:  Medchemexpress LLC HY10001125MG
Encompass_Preferred
SR-3029 is a potent, ATP-competitive inhibitor of casein kinase 1 delta (CK1δ) and epsilon (CK1ε) with characterized biochemical, cellular, and in vivo activity. It is used in kinase signaling and oncology research to probe CK1-dependent pathways and evaluate antiproliferative effects in cell and animal models.
  • Potent ATP-competitive inhibitor of CK1δ and CK1ε (IC50s 44 nM and 260 nM).
  • Ki of 97 nM against CK1 isoforms.
  • Cellular antiproliferative activity with EC50s in the low nanomolar range.
  • Demonstrated antitumor activity in multiple xenograft models.
  • High reported purity (99.73%).
  • Available as solid and solution formats for biochemical and cell-based assays.
Catalog No. 50-002-15665
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.