Learn More
Medchemexpress LLC N-((6,7-difluoro-1H-benzo[d]imidazol-2-yl)methyl)-9-(3-fluorophenyl)-2-morpholino-9H-purin-6-amine | 1454585-06-8 | MFCD30187527 | 99.7% | 480.45 | C23H19F3N8O | 5 MG
Supplier: Medchemexpress LLC HY1000115MG
SR-3029 is a small-molecule, ATP-competitive inhibitor selective for casein kinase 1 delta and epsilon (CK1δ/CK1ε). It is used as a research tool compound to study CK1-dependent signaling pathways and to evaluate antiproliferative effects in preclinical models.
- Selective CK1δ/CK1ε inhibition with IC50 values in the low-nanomolar range.
- ATP-competitive mechanism useful for kinase-targeting studies.
- High chemical purity (99.73%).
- Supplied as a solid suitable for formulation and in vitro assays.
- Well-characterized structural identifiers (CAS, SMILES, MFCD) for unambiguous identification.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.