Learn More
Medchemexpress LLC SR-4370 | 1816294-67-3 | 98.38% | 304.33 | 500 MG
SDP
Supplier:  Medchemexpress LLC HY111400100MG
Encompass_Preferred
SR-4370 is an inhibitor of Histone Deacetylase (HDAC) and exhibits cytotoxicity against breast cancer cells.
  • Inhibits HDAC1, HDAC2, HDAC3, HDAC8, and HDAC6.
  • Cytotoxic to MDA-MB-231 breast cancer cells.
Catalog No. 50-003-43373