Learn More
TARGETMOL CHEMICALS INC SR-4835 5MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T83255MG
Encompass_Preferred
Also available in 1 mg 2 mg 10 mg 25 mg 50 mg 100 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. SR-4835 is a highly selective dual inhibitor of CDK12 and CDK13(CDK12 IC50=99 nM Kd=98 nM; CDK13 Kd=4.9 nM) which disables triple-negative breast cancer (TNBC) cellspurity: 99%
Catalog No. 50-257-6248