Learn More
Abcam SR 59230A hydrochloride, beta3 adrenoceptor antagonist, 10MG
Supplier:  Abcam AB14113510MG
Encompass_Preferred
MW 361.9 Da, Purity >99%. Potent, selective β3 adrenoceptor antagonist (IC50 values are 40, 408 and 648 nM for β3, β1 and β2, respectively). Orally active *in vivo*.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-6862