Learn More
Medchemexpress LLC SR-717 free acid | 2375420-34-9 | 99.4% | 345.26 | C15H9F2N5O3 | 5 MG

Supplier: Medchemexpress LLC HY131454A5MG
SR-717 free acid is a non-nucleotide STING agonist provided as a high-purity research compound for cellular assays that probe STING-mediated immune responses. The compound includes documented solubility and storage recommendations for laboratory use.
- High purity of 99.36%.
- Molecular formula C15H9F2N5O3; molecular weight 345.26.
- CAS number 2375420-34-9.
- Cellular activity: EC50 2.1 μM (ISG-THP1 WT) and 2.2 μM (ISG-THP1 cGAS KO).
- Solubility in water 6.11 mg/mL (adjust pH to 12 with 1 M NaOH).
- Storage: powder -20°C (long term) or 4°C (short term); solutions -80°C.
- Supplied in small research quantities, including 5 mg.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.