Learn More
ABclonal Technology SR-BI Rabbit pAb
SDP
Supplier:  ABclonal Technology A1584500UL
Encompass_Preferred
The protein encoded by this gene is a plasma membrane receptor for high density lipoprotein cholesterol (HDL). The encoded protein mediates cholesterol transfer to and from HDL. In addition, this protein is a receptor for hepatitis C virus glycoprotein E2 and facilitates cell entry by the virus, SARS-CoV2.
Catalog No. 50-001-08700