Learn More
Medchemexpress LLC SR-BI/SCARB1 protein, human (recombinant, C-terminal His tag) | >95.0% | 10 UG

Supplier: Medchemexpress LLC HYP7665710UG
Recombinant human SR-BI/SCARB1 extracellular domain expressed in HEK293 cells, supplied C-terminal His-tagged and lyophilized for biochemical and ligand-binding research. Supplied as a 10 μg vial with >95% purity and formulated from PBS with stabilizing excipients; recommended storage at -20°C or -80°C for long-term preservation.
- Recombinant human extracellular domain expressed in HEK293 cells.
- C-terminal His tag for affinity purification and detection.
- Amino acid range P33-Y443 representing the mature extracellular sequence.
- Purity >95.0% by reducing SDS-PAGE.
- Lyophilized formulation with trehalose or mannitol and 0.01% Tween 80 for stability.
- Suitable for ligand binding, receptor interaction, and antibody validation studies.
- Store at -20°C; aliquot and freeze at -80°C for long-term storage.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.