Learn More
Eichrom Industries SR Resin Cartridge, 2 mL, Package of 50, 40% Loading, 50-100 μm Particle Size, Maximum Capacity 12 mg Sr, Recommended Working Capacity 8 mg Sr, Inert Polymeric Support
SDP
Supplier:  Eichrom Industries SRR50S
Encompass
Eichrom Technologies offers a solution for strontium separation via specialized cartridges that enhance efficiency in scientific applications. These cartridges are designed to work effectively with high nitric acid concentrations, allowing for reliable isolation from complex matrices. They are essential in radiochemistry and environmental testing, where precise strontium analysis is vital.
  • Highly selective extractant system tailored for strontium
  • Convenient 2 mL cartridge format
  • Minimal interference from calcium and sodium elements
  • Supports efficient flow rates with specialized particle size
  • Recommended working capacity of 8 mg Sr per cartridge
  • Easy elution with dilute HNO3 or deionized water
  • Approximately 90 Sr k' value at 8M HNO3
Catalog No. NC1553099