Learn More
Medchemexpress LLC SR11237 100mg | 146670-40-8 | 100MG
SDP
Supplier:  Medchemexpress LLC HY107413100MG
Encompass_Preferred
SR11237 (BMS-649) is a potent retinoid X receptor (RXR)-selective agonist that is devoid of any RAR activity SR11237 can cause RXR/RXR homodimers to form and transactivate a reporter gene containing a RXR-response element[1 [2 [3
Catalog No. 50-002-39720