Learn More
Medchemexpress LLC HY-103200 5mg Medchemexpress, SR59230A (hydrochloride) CAS:1135278-41-9 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1032005MG
Encompass_Preferred
Medchemexpress, HY-103200 5mg SR59230A (hydrochloride) CAS:1135278-41-9 SR59230A hydrochloride is a potent, selective, and blood-brain barrier penetrating β3-adrenergic receptor antagonist with IC50s of 40, 408, and 648 nM for β3, β1, and β2 receptors, respectively. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-0659