Learn More
Medchemexpress LLC Src-3-in-2 (SI-12 6c) | 3074959-44-4 | 96.3% | 319.28 | C15H12F3N5 | 10 MG

Supplier: Medchemexpress LLC HY16346910MG
SRC-3-IN-2 (SI-12 6c) is an orally active inhibitor of steroid receptor coactivator 3 (SRC-3) with reported antitumor activity. It is supplied as a research-grade solid and as a DMSO solution for biochemical and cellular studies.
- Orally active SRC-3 inhibitor with reported antitumor activity.
- Suitable for biochemical and cellular assays.
- Available as a solid and 10 mM solution in DMSO.
- Purity approximately 96.3%.
- Molecular weight 319.28; formula C15H12F3N5.
- Supplied in small research pack sizes, including 10 mg.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.