Learn More
Enzo Life Sciences Src I1 (25mg). CAS: 179248-59-0
SDP
Supplier:  Enzo Life Sciences BMLEI3980025
Encompass_Preferred
Potent, dual site (ATP domain and substrate-binding domain) Src and Lck kinase inhibitor (IC50=44 and 88nM respectively). VEGFR2 IC50=0.32μM; c-fms IC50=30μM. Alternative name: 6,7-Dimethoxy-N-(4-phenoxyphenyl)-4-quinazolinamine. Purity: ≥98% (HPLC). Solubility: Soluble in DMSO (5mg/ml). Long Term Storage: Ambient.
Catalog No. 50-201-0778