Learn More
Medchemexpress LLC Src inhibitor 1 | 179248-59-0 | MFCD01815300 | 100.0% | 373.40 | C22H19N3O3 | 50 MG
SDP
Supplier:  Medchemexpress LLC HY10105350MG
Encompass_Preferred
Src Inhibitor 1 is a potent, ATP-competitive dual-site inhibitor of Src-family tyrosine kinases with reported IC50 values of 44 nM for Src and 88 nM for Lck. Supplied as a high-purity research chemical, it is intended for biochemical and cellular studies probing Src-family kinase signaling.
  • Potent ATP-competitive inhibition of Src-family kinases.
  • Dual-site binding profile enabling effective kinase blockade.
  • Reported IC50 values: Src 44 nM, Lck 88 nM.
  • High purity suitable for research applications.
  • Available in multiple pack sizes, including 50 mg solid form.
  • Appropriate for biochemical and cellular assay use.
Catalog No. 50-002-12263