Learn More
Cayman Chemical Src KInase InhIbItr I 10mg
SDP
Supplier:  Cayman Chemical 1459210 MG
Encompass_Preferred
A potent competitive inhibitor of both Src and Lck (IC50 = 44 and 88 nM, respectively), as well as Csk and Yes; also inhibits RIP-2
Catalog No. 50-297-2687