Learn More
TARGETMOL CHEMICALS INC SRI-011381 10MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T428310MG
Encompass_Preferred
Also available in 1 mL, 1 mg, 2 mg, 5 mg, 25 mg, 50 mg, 100 mg, 200 mg, 500 mg and bulk. Please contact Fisher for quotes. SRI-011381, a new-type agonist of the TGF-beta signaling pathway, is utilized in the treatment of Alzheimer's disease. Purity 99.11%
Catalog No. 50-287-8953