Learn More
Medchemexpress LLC SRI-011381 hydrochloride | 2070014-88-7 | 99.6% | 365.94 g/mol | C20H32ClN3O | 1 G
SDP
Supplier:  Medchemexpress LLC HY100347A1G
Encompass_Preferred
SRI-011381 hydrochloride is a small-molecule, orally active agonist of the TGF-β/Smad signaling pathway with reported neuroprotective effects. Supplied as the hydrochloride salt for research use, the compound is characterized by a defined molecular weight and high reported purity suitable for in vitro and in vivo studies.
  • Orally active TGF-β signaling agonist.
  • Reported neuroprotective effects in preclinical studies.
  • Supplied as the hydrochloride salt for improved handling and solubility.
  • High reported purity (99.55%).
  • Molecular weight 365.94 g/mol.
  • For research use only.
Catalog No. 50-002-14398