Learn More
Medchemexpress LLC SRI-011381 hydrochloride | 2070014-88-7 | 99.6% | 365.94 g/mol | C20H32ClN3O | 10 MG
SDP
Supplier:  Medchemexpress LLC HY100347A10MG
Encompass_Preferred
SRI-011381 hydrochloride is an orally active TGF-β signaling agonist used in research. It promotes fibroblast proliferation and increases TGF-β1, NALP3, collagen-1, and α-SMA expression in vitro, and reduces neuronal loss and inflammatory infiltration in animal models.
  • Orally active TGF-β signaling agonist for in vivo and in vitro studies.
  • Promotes fibroblast proliferation and extracellular matrix marker expression.
  • Demonstrated neuroprotective effects in animal disease models.
  • High purity suitable for research applications.
  • Soluble in DMSO, water, and PBS with ultrasonic assistance.
Catalog No. 50-002-14371