Learn More
Medchemexpress LLC SRI-37330 hydrochloride | 2322245-49-6 | 99.7% | C16H20ClF3N4O2S | 1 MG
SDP
Supplier:  Medchemexpress LLC HY1416231MG
Encompass_Preferred
SRI-37330 hydrochloride is an orally bioavailable thioredoxin-interacting protein (TXNIP) inhibitor. It inhibits glucagon secretion and function, reduces hepatic glucose production, and reverses hepatic steatosis. This compound can be used for type 2 diabetes research.
  • Orally bioavailable thioredoxin-interacting protein (TXNIP) inhibitor
  • Inhibits glucagon secretion and function
  • Reduces hepatic glucose production
  • Reverses hepatic steatosis
  • Used for type 2 diabetes research
Catalog No. 50-003-53883