Learn More
Medchemexpress LLC SRI-37330 hydrochloride | 2322245-49-6 | 99.7% | C16H20ClF3N4O2S | 100 MG
SDP
Supplier:  Medchemexpress LLC HY141623100MG
Encompass_Preferred
SRI-37330 hydrochloride is an orally bioavailable thioredoxin-interacting protein (TXNIP) inhibitor. It inhibits glucagon secretion and function, reduces hepatic glucose production, and reverses hepatic steatosis. This compound is used for type 2 diabetes research.
  • Inhibits human TXNIP promoter activity in INS-1 cells
  • Reduces TXNIP mRNA and protein levels in INS-1 cells
  • Inhibits Pol II binding to the E-box motif of the TXNIP promoter
  • Lowers glucagon secretion in TC1-6 cells
  • Inhibits glucagon-induced glucose output from primary hepatocytes
  • Decreases glucagon secretion and blocks hepatic glucose output in mice
  • Well tolerated in C57BL/6J mice
  • Reverses obesity- and streptozotocin-induced diabetes and hepatic steatosis in mice
Catalog No. 50-003-56111