Learn More
Medchemexpress LLC SRI-37330 hydrochloride | 2322245-49-6 | 99.7% | C16H20ClF3N4O2S | 50 MG
SDP
Supplier:  Medchemexpress LLC HY14162350MG
Encompass_Preferred
SRI-37330 hydrochloride is an orally bioavailable thioredoxin-interacting protein (TXNIP) inhibitor that reduces glucagon secretion and hepatic glucose production, and reverses hepatic steatosis. It can be used for type 2 diabetes research.
  • Inhibits activity of human TXNIP promoter in INS-1 cells
  • Lowers mRNA and protein levels of TXNIP
  • Inhibits polymerase II (Pol II) binding to E-box motif region
  • Lowers glucagon secretion in TC1-6 cells
  • Inhibits glucagon-induced glucose output from primary hepatocytes
  • Decreases glucagon secretion and action in male C57BL/6J mice
  • Blocks hepatic glucose output in male C57BL/6J mice
  • Reverses obesity- and Streptozotocin (STZ)-induced diabetes and hepatic steatosis in mice
Catalog No. 50-003-55982