Learn More
Selleck Chemical LLC SRPIN340
SDP
Supplier:  Selleck Chemical LLC S727050MG
Encompass_Preferred
SRPIN340 is a selective SRPK inhibitor with Ki of 0 89 M for SRPK1 showing no significant inhibitory activity against more than 140 other kinases
Catalog No. 50-000-03502