Learn More
Medchemexpress LLC HY-131959 100mg Medchemexpress, (S,R,S)-AHPC-PEG5-Boc CAS: Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY131959100MG
Encompass_Preferred
Medchemexpress, HY-131959 100mg (S,R,S)-AHPC-PEG5-Boc CAS: (S,R,S)-AHPC-PEG5-Boc is a E3 ligase ligand-linker conjugate that incorporates the (S,R,S)-AHPC based VHL ligand and a linker used for Cdc20 degrader CP5V. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-1301