Learn More
Medchemexpress LLC SRT 1460 | 925432-73-1 | >98.0% | 507.60 g·mol⁻¹ | C26H29N5O4S | 5MG
SDP
Supplier:  Medchemexpress LLC HY1240375MG
Encompass_Preferred
SRT 1460 is a small-molecule activator of sirtuin 1 (SIRT1) used in biochemical and cellular research. It shows potent activation of SIRT1 (reported EC1.5 ≈ 2.9 μM), with selectivity versus SIRT2 and SIRT3. The compound is supplied as a high-purity research reagent (CAS 925432-73-1), chemical formula C26H29N5O4S, and molecular weight 507.60 g·mol⁻¹.
Catalog No. 50-002-37180