Learn More
Selleck Chemical LLC SRT2104 (GSK2245840) 5mg 1093403-33-8
SDP
Supplier:  Selleck Chemical LLC S77925MG
Encompass_Preferred
SRT2104 (GSK2245840) is a selective SIRT1 activator involved in the regulation of energy homeostasis. Phase 2. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-136-5253