Learn More
Medchemexpress LLC Srt3109 | 1204707-71-0 | C18H23F2N5O4S2 | 100 MG
SDP
Supplier:  Medchemexpress LLC HY15462100MG
Encompass_Preferred
SRT3109 is an antagonist of CXCR2, with a pIC50 of 8.2, and is utilized in the research of chemokine-mediated diseases. It is intended for research use only and is not sold to patients. The product appears as a solid with a white to off-white color and has a molecular weight of 475.53 and a purity of 99.68%.
  • Antagonist of CXCR2
  • Used in research of chemokine mediated diseases
  • Appears as a white to off-white solid
  • High purity of 99.68%
  • Molecular weight of 475.53
Catalog No. 50-003-76675